siRNA Details
| Gene Symbol: DISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020780.1 | NP_065831.1 | 21 | 5154 | ||
| XM_011541828.2 | XP_011540130.1 | 20 | 7256 | |||
| XM_011541829.1 | XP_011540131.1 | 20 | 4129 | |||
| GenBank mRNA | AB037758.1 | - | 21 | 5154 | ||
| AB593117.1 | - | 21 | 5237 | |||

DISP3
Genomic Map