siRNA Details
| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_020823.1 | NP_065874.1 | 5, 6 | 809 | ||
| XM_005267074.3 | XP_005267131.1 | 5, 6 | 526 | |||
| XM_011536002.2 | XP_011534304.1 | 5, 6 | 991 | |||
| GenBank mRNA | AB037844.1 | - | 5, 6 | 811 | ||
| AY927471.1 | - | LIT3489 AY927471 |
5, 6 | 926 | ||
| AY927472.1 | - | 6, 7 | 1462 | |||
| BC033450.1 | - | 4, 5 | 532 | |||

TMEM181
Genomic Map