siRNA Details
Gene Symbol: SLAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020846.1 | NP_065897.1 | 8 | 3422 | ||
XM_005248121.2 | XP_005248178.1 | 9 | 3616 | |||
GenBank mRNA | AB040891.1 | - | 8 | 3392 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020846.1 | NP_065897.1 | 8 | 3422 | ||
XM_005248121.2 | XP_005248178.1 | 9 | 3616 | |||
GenBank mRNA | AB040891.1 | - | 8 | 3392 |