siRNA Details
| Gene Symbol: KIAA1462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020848.2 | NP_065899.1 | 4 | 6203 | ||
| XM_011519608.1 | XP_011517910.1 | 4 | 6242 | |||
| XM_011519609.2 | XP_011517911.1 | 5 | 6270 | |||
| GenBank mRNA | AB040895.2 | - | 3 | 6097 | ||
| BC047548.1 | - | 2 | 3516 | |||
| BC071601.1 | - | 3 | 2440 | |||
| BX648263.1 | - | 2 | 5123 | |||

KIAA1462
Genomic Map