siRNA Details
| Gene Symbol: LRFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020862.1 | NP_065913.1 | 1 | 1373 | ||
| XM_005259102.4 | XP_005259159.1 | 4 | 1573 | |||
| XM_017027033.1 | XP_016882522.1 | 4 | 1688 | |||
| GenBank mRNA | AB040917.1 | - | 1 | 1162 | ||
| BC014678.1 | - | 469 | ||||
| BC025310.1 | - | 1 | 686 | |||

LRFN1
Genomic Map