siRNA Details
| Gene Symbol: NYAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020864.1 | NP_065915.1 | 6 | 2908 | ||
| XM_005246708.3 | XP_005246765.1 | 7 | 3779 | |||
| GenBank mRNA | AB040919.1 | - | 6 | 2816 | ||
| BC131727.1 | - | 4 | 1441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NYAP2| Gene Symbol: NYAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020864.1 | NP_065915.1 | 6 | 2908 | ||
| XM_005246708.3 | XP_005246765.1 | 7 | 3779 | |||
| GenBank mRNA | AB040919.1 | - | 6 | 2816 | ||
| BC131727.1 | - | 4 | 1441 | |||