siRNA Details
| Gene Symbol: KIAA1549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164665.1 | NP_001158137.1 | 20 | 6445 | ||
| NM_020910.2 | NP_065961.2 | 20 | 6397 | |||
| XM_011516442.2 | XP_011514744.1 | 21 | 6672 | |||
| GenBank mRNA | AB046769.2 | - | 19 | 6141 | ||
| AL136736.1 | - | 3 | 920 | |||
| BC038232.1 | - | 15 | 3026 | |||
| BC091523.1 | - | 5 | 1355 | |||

KIAA1549
Genomic Map