siRNA Details
| Gene Symbol: ZSWIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020928.1 | NP_065979.1 | 2 | 863 | ||
| GenBank mRNA | BF750632.1 | - | 1 | 83 | ||
| BF751445.1 | - | 1 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSWIM6| Gene Symbol: ZSWIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020928.1 | NP_065979.1 | 2 | 863 | ||
| GenBank mRNA | BF750632.1 | - | 1 | 83 | ||
| BF751445.1 | - | 1 | 90 | |||