siRNA Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037806.3 | NP_001032895.2 | 8 | 2877 | ||
| XM_005269050.3 | XP_005269107.1 | 8 | 3202 | |||
| XM_006719525.2 | XP_006719588.1 | 8 | 3172 | |||
| XM_011538610.2 | XP_011536912.1 | 9 | 2956 | |||
| XM_011538611.2 | XP_011536913.1 | 9 | 2926 | |||
| GenBank mRNA | AB046822.1 | - | 1 | 1818 | ||
| AF370366.1 | - | 1 | 185 | |||
| BC007998.1 | - | 2 | 396 | |||
| BC033253.1 | - | 1 | 1393 | |||

NCKAP5L
Genomic Map