siRNA Details
| Gene Symbol: PLEKHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021623.1 | NP_067636.1 | 7 | 784 | ||
| XM_011544605.2 | XP_011542907.1 | 7 | 669 | |||
| XM_011544606.2 | XP_011542908.1 | 7 | 720 | |||
| XM_011544607.2 | XP_011542909.1 | 7 | 682 | |||
| XM_017013719.1 | XP_016869208.1 | 6 | 505 | |||
| XM_017013720.1 | XP_016869209.1 | 6 | 620 | |||
| GenBank mRNA | AF286164.1 | - | 7 | 709 | ||
| AK098787.1 | - | 7 | 785 | |||
| AK291842.1 | - | 7 | 728 | |||
| AK296068.1 | - | 6 | 577 | |||

PLEKHA2
Genomic Map