siRNA Details
| Gene Symbol: PEAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024776.3 | NP_079052.2 | 5 | 1225 | ||
| XM_005254670.2 | XP_005254727.1 | 8 | 1529 | |||
| XM_005254671.2 | XP_005254728.1 | 7 | 1426 | |||
| XM_005254673.2 | XP_005254730.1 | 5 | 1160 | |||
| XM_005254674.2 | XP_005254731.1 | 6 | 1373 | |||
| XM_005254675.4 | XP_005254732.1 | 2 | 879 | |||
| XM_011522033.2 | XP_011520335.1 | 9 | 1585 | |||
| XM_011522034.2 | XP_011520336.1 | 10 | 1693 | |||
| XM_011522035.2 | XP_011520337.1 | 12 | 1844 | |||
| XM_011522036.2 | XP_011520338.1 | 11 | 1748 | |||
| XM_011522037.2 | XP_011520339.1 | 11 | 1789 | |||
| XM_011522038.2 | XP_011520340.1 | 10 | 1640 | |||
| XM_011522039.2 | XP_011520341.1 | 11 | 1736 | |||
| XM_011522040.1 | XP_011520342.1 | 9 | 1585 | |||
| XM_011522041.1 | XP_011520343.1 | 9 | 1585 | |||

PEAK1
Genomic Map