siRNA Details
| Gene Symbol: ASXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030632.2 | NP_085135.1 | 11 | 2527 | ||
| XM_005258356.1 | XP_005258413.1 | 12 | 2500 | |||
| XM_011526205.2 | XP_011524507.1 | 12 | 9290 | |||
| XM_011526206.2 | XP_011524508.1 | 12 | 6055 | |||
| XM_011526209.1 | XP_011524511.1 | 13 | 2684 | |||
| XM_011526210.1 | XP_011524512.1 | 13 | 2741 | |||
| XM_011526212.1 | XP_011524514.1 | 12 | 2645 | |||
| XM_011526213.1 | XP_011524515.1 | 12 | 2738 | |||
| XM_017026012.1 | XP_016881501.1 | 12 | 5902 | |||
| XM_017026013.1 | XP_016881502.1 | 13 | 6264 | |||
| XM_017026014.1 | XP_016881503.1 | 13 | 2701 | |||
| GenBank mRNA | AB051500.2 | - | 1 | 686 | ||
| AK131454.1 | - | 13 | 2570 | |||
| AL133050.1 | - | 1 | 22 | |||

ASXL3
Genomic Map