siRNA Details
| Gene Symbol: KDM7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030647.1 | NP_085150.1 | 12, 13 | 1650 | ||
| XM_011516587.2 | XP_011514889.1 | 6, 7 | 675 | |||
| GenBank mRNA | AK131497.1 | - | 12, 13 | 1651 | ||
| BX641017.1 | - | 4, 5 | 430 | |||
| BX649115.1 | - | 7, 8 | 917 | |||

KDM7A
Genomic Map