siRNA Details
| Gene Symbol: FRMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032428.1 | NP_115804.1 | 10 | 894 | ||
| XM_006724709.2 | XP_006724772.1 | 4 | 369 | |||
| XM_011531056.2 | XP_011529358.1 | 8 | 870 | |||
| XM_011531057.2 | XP_011529359.1 | 8 | 796 | |||
| XM_011531058.2 | XP_011529360.1 | 7 | 708 | |||
| XM_017029901.1 | XP_016885390.1 | 8 | 1115 | |||
| XM_017029902.1 | XP_016885391.1 | 8 | 1115 | |||

FRMPD3
Genomic Map