siRNA Details
| Gene Symbol: TMEM132B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052907.3 | NP_443139.2 | 2 | 187 | ||
| XM_011537852.2 | XP_011536154.1 | 2 | 244 | |||
| XM_017018767.1 | XP_016874256.1 | 3 | 353 | |||
| GenBank mRNA | CR735824.1 | - | 2 | 188 | ||

TMEM132B
Genomic Map