siRNA Details
| Gene Symbol: ZNF594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032530.1 | NP_115919.1 | 2 | 4114 | ||
| XM_005256827.2 | XP_005256884.1 | 2 | 4427 | |||
| GenBank mRNA | AB058774.2 | - | 1 | 4099 | ||
| AK131271.1 | - | 3 | 3286 | |||
| AL833185.1 | - | 2 | 2633 | |||
| BC136604.1 | - | 1 | 4096 | 9020594 | ||

ZNF594
Genomic Map