siRNA Details
Gene Symbol: KIAA1644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099294.1 | NP_001092764.1 | 5 | 5872 | ||
XM_005261790.3 | XP_005261847.1 | 5 | 5970 | |||
GenBank mRNA | AB051431.1 | - | 3 | 5571 | ||
BC068280.1 | - | 1 | 4458 |
