siRNA Details
| Gene Symbol: ZNF479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033273.2 | NP_150376.1 | 5 | 1024 | ||
| XM_011515604.2 | XP_011513906.1 | 6 | 1210 | |||
| XM_011515608.2 | XP_011513910.1 | 6 | 1114 | |||
| XM_011515609.2 | XP_011513911.1 | 4 | 1005 | |||
| XM_011515610.2 | XP_011513912.1 | 7 | 1291 | |||
| XM_011515611.2 | XP_011513913.1 | 5 | 1158 | |||
| XM_017012777.1 | XP_016868266.1 | 4 | 859 | |||
| XM_017012778.1 | XP_016868267.1 | 5 | 1062 | |||
| GenBank mRNA | AF277624.1 | - | 5 | 1024 | ||

ZNF479
Genomic Map