siRNA Details
| Gene Symbol: ZNF251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138367.1 | NP_612376.1 | 5 | 2961 | ||
| GenBank mRNA | BC006258.2 | - | 1 | 1572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF251| Gene Symbol: ZNF251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138367.1 | NP_612376.1 | 5 | 2961 | ||
| GenBank mRNA | BC006258.2 | - | 1 | 1572 | ||