siRNA Details
| Gene Symbol: ZNF845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321522.1 | NP_001308451.1 | 5 | 3515 | ||
| NM_001321523.1 | NP_001308452.1 | 5 | 3454 | |||
| NM_001321524.1 | NP_001308453.1 | 5 | 3462 | |||
| NM_138374.2 | NP_612383.1 | 4 | 3360 | |||
| GenBank mRNA | AK096653.1 | - | 1 | 1783 | ||
| AK295079.1 | - | 6 | 3182 | |||
| BC007307.1 | - | 1 | 2216 | |||

ZNF845
Genomic Map