siRNA Details
| Gene Symbol: RIPPLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171706.1 | NP_001165177.1 | 2 | 1013 | ||
| NM_138382.2 | NP_612391.1 | 4 | 1154 | |||
| GenBank mRNA | BC007652.1 | - | 4 | 1140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIPPLY1| Gene Symbol: RIPPLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171706.1 | NP_001165177.1 | 2 | 1013 | ||
| NM_138382.2 | NP_612391.1 | 4 | 1154 | |||
| GenBank mRNA | BC007652.1 | - | 4 | 1140 | ||