siRNA Details
| Gene Symbol: DUSP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080426.1 | NP_001073895.1 | 5 | 2886 | ||
| XM_005245603.3 | XP_005245660.1 | 6 | 3049 | |||
| XM_011510146.2 | XP_011508448.1 | 5 | 2923 | |||
| GenBank mRNA | AF119045.1 | - | 1 | 1395 | ||
| AK094814.1 | - | 1 | 1483 | |||
| BC115387.1 | - | 6 | 3006 | |||

DUSP27
Genomic Map