siRNA Details
| Gene Symbol: TMEM132C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136103.2 | NP_001129575.2 | 8 | 1984 | ||
| XM_011538998.2 | XP_011537300.1 | 8 | 2310 | |||
| GenBank mRNA | AK126715.1 | - | 5 | 853 | ||
| AL831921.1 | - | 5 | 831 | |||
| AL832160.1 | - | 3 | 458 | |||

TMEM132C
Genomic Map