siRNA Details
| Gene Symbol: TMEM132C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136103.2 | NP_001129575.2 | 9 | 4169 | ||
| XM_011538998.2 | XP_011537300.1 | 9 | 4495 | |||
| GenBank mRNA | AK126715.1 | - | 6 | 3038 | ||
| AL122107.1 | - | 1 | 1252 | |||
| AL831921.1 | - | 6 | 3016 | |||
| AL832160.1 | - | 4 | 2643 | |||

TMEM132C
Genomic Map