siRNA Details
| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195304.1 | NP_001182233.1 | 2, 3 | 347 | ||
| GenBank mRNA | AK021501.1 | - | 2, 3 | 148 | ||
| AK307314.1 | - | 2, 3 | 223 | |||
| BC105804.1 | - | 2, 3 | 186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BBIP1| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195304.1 | NP_001182233.1 | 2, 3 | 347 | ||
| GenBank mRNA | AK021501.1 | - | 2, 3 | 148 | ||
| AK307314.1 | - | 2, 3 | 223 | |||
| BC105804.1 | - | 2, 3 | 186 | |||