siRNA Details
| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195304.1 | NP_001182233.1 | 5 | 722 | ||
| NM_001195305.1 | NP_001182234.1 | 4 | 565 | |||
| NM_001195306.1 | NP_001182235.1 | 4 | 411 | |||
| NM_001195307.1 | NP_001182236.1 | 3 | 490 | |||
| NM_001243783.1 | NP_001230712.1 | 3 | 472 | |||
| GenBank mRNA | AK025724.1 | - | 4 | 385 | ||
| AK125829.1 | - | 1 | 4569 | |||
| AK307126.1 | - | 4 | 411 | |||
| AK307314.1 | - | 5 | 598 | |||
| BC105804.1 | - | 5 | 561 | |||
| BI552279.1 | - | 3 | 304 | |||

BBIP1
Genomic Map