siRNA Details
| Gene Symbol: TRANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014831.2 | NP_055646.2 | 11, 12 | 1796 | ||
| XM_006713432.3 | XP_006713495.1 | 12, 13 | 1964 | |||
| XM_011534289.2 | XP_011532591.1 | 12, 13 | 2725 | |||
| XM_017007570.1 | XP_016863059.1 | 11, 12 | 1596 | |||
| XM_017007571.1 | XP_016863060.1 | 12, 13 | 1857 | |||
| XM_017007572.1 | XP_016863061.1 | 12, 13 | 1759 | |||
| XM_017007573.1 | XP_016863062.1 | 12, 13 | 1767 | |||
| GenBank mRNA | AB002340.3 | - | 1, 2 | 174 | ||
| BG750754.1 | - | 1, 2 | 200 | |||

TRANK1
Genomic Map