siRNA Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128127.2 | NP_001121599.1 | 19, 20 | 1823 | ||
| NM_001205019.1 | NP_001191948.1 | 20, 21 | 1841 | |||
| XM_005274488.4 | XP_005274545.1 | 17, 18 | 1593 | |||
| XM_006724483.2 | XP_006724546.1 | 20, 21 | 1860 | |||
| XM_006724485.2 | XP_006724548.1 | 19, 20 | 1686 | |||
| XM_006724486.3 | XP_006724549.1 | 18, 19 | 1671 | |||
| XM_011545491.2 | XP_011543793.1 | 21, 22 | 1879 | |||
| XM_011545493.2 | XP_011543795.1 | 19, 20 | 1561 | |||
| XM_011545494.2 | XP_011543796.1 | 19, 20 | 1506 | |||
| XM_017029409.1 | XP_016884898.1 | 21, 22 | 1876 | |||
| GenBank mRNA | AK294938.1 | - | 21, 22 | 1876 | ||
| AK308238.1 | - | 17, 18 | 1503 | |||
| Gene Symbol: GK3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026575.1 | 1 | 2022 | |||
| GenBank mRNA | AK225740.1 | - | 1 | 1673 | ||
| AK292282.1 | - | 1 | 2022 | |||
| BC066960.1 | - | 1 | 1678 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001294140, NM_212444, CF740682, AK088373, AK044308.

GK
Genomic Map