siRNA Details
| Gene Symbol: Ccser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164316.1 | NP_001157788.1 | 11 | 2696 | ||
| NM_183310.2 | NP_899133.2 | 11 | 2748 | |||
| GenBank mRNA | AK173231.1 | - | 10 | 1441 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccser1| Gene Symbol: Ccser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164316.1 | NP_001157788.1 | 11 | 2696 | ||
| NM_183310.2 | NP_899133.2 | 11 | 2748 | |||
| GenBank mRNA | AK173231.1 | - | 10 | 1441 | ||