siRNA Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085471.1 | NP_001078940.1 | 7 | 7286 | ||
| NM_005197.3 | NP_005188.2 | 6 | 7337 | |||
| GenBank mRNA | AF116643.2 | - | 1 | 1211 | ||
| AK055175.1 | - | 2 | 2954 | |||
| AK123929.1 | - | 1 | 1962 | |||
| BC010460.1 | - | 1 | 1077 | |||

FOXN3
Genomic Map