siRNA Details
| Gene Symbol: PLXNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012401.3 | NP_036533.2 | 12 | 2318 | ||
| XM_005261909.1 | XP_005261966.1 | 13 | 2366 | |||
| XM_005261910.2 | XP_005261967.1 | 13 | 2377 | |||
| XM_005261911.4 | XP_005261968.1 | 12 | 2316 | |||
| XM_006724413.2 | XP_006724476.1 | 14 | 2436 | |||
| XM_011530682.2 | XP_011528984.1 | 13 | 2517 | |||
| XM_017028703.1 | XP_016884192.1 | 14 | 2417 | |||
| XM_017028704.1 | XP_016884193.1 | 13 | 2370 | |||
| GenBank mRNA | AB002313.1 | - | 11 | 2235 | ||

PLXNB2
Genomic Map