siRNA Details
| Gene Symbol: ZNF275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080485.3 | NP_001073954.3 | 5 | 2092 | ||
| GenBank mRNA | AK122976.1 | - | 5 | 2092 | ||
| AK127133.1 | - | 1 | 1888 | |||
| BC136634.1 | - | 2 | 3216 | 9020624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF275| Gene Symbol: ZNF275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080485.3 | NP_001073954.3 | 5 | 2092 | ||
| GenBank mRNA | AK122976.1 | - | 5 | 2092 | ||
| AK127133.1 | - | 1 | 1888 | |||
| BC136634.1 | - | 2 | 3216 | 9020624 | ||