siRNA Details
| Gene Symbol: VPS37D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077621.1 | NP_001071089.1 | 4 | 1526 | ||
| XM_017011779.1 | XP_016867268.1 | 4 | 1716 | |||
| GenBank mRNA | AY081952.1 | - | 4 | 1526 | ||
| BC064621.1 | - | 4 | 1373 | |||

VPS37D
Genomic Map