siRNA Details
| Gene Symbol: OVOS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020288.1 | XP_016875777.1 | 2 | 104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OVOS2| Gene Symbol: OVOS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020288.1 | XP_016875777.1 | 2 | 104 | ||