siRNA Details
| Gene Symbol: LINC00342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057596.1 | - | 1 | 480 | ||
| BC117383.1 | - | 1 | 450 | |||
| BC126153.1 | - | 1 | 450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00342| Gene Symbol: LINC00342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057596.1 | - | 1 | 480 | ||
| BC117383.1 | - | 1 | 450 | |||
| BC126153.1 | - | 1 | 450 | |||