siRNA Details
Gene Symbol: LOC100130417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026874.2 | 4 | 720 | |||
NR_122045.1 | 4 | 660 | ||||
GenBank mRNA | AK096928.1 | - | 1 | 3413 | ||
BC025320.1 | - | 1 | 1067 | |||
BC144023.1 | - | 4 | 465 | |||
BC144024.1 | - | 4 | 405 |
