siRNA Details
| Gene Symbol: VWA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173615.4 | NP_775886.3 | 33 | 3643 | ||
| XM_011545742.2 | XP_011544044.1 | 37 | 3925 | |||
| XM_011545744.2 | XP_011544046.1 | 22 | 2525 | |||
| XM_011545745.1 | XP_011544047.1 | 20 | 2293 | |||
| XM_011545746.2 | XP_011544048.1 | 20 | 2396 | |||
| XM_017022947.1 | XP_016878436.1 | 38 | 3998 | |||
| XM_017022948.1 | XP_016878437.1 | 21 | 3097 | |||
| XM_017022949.1 | XP_016878438.1 | 19 | 3368 | |||
| GenBank mRNA | AK093467.1 | - | 9 | 1206 | ||
| AK128606.1 | - | 22 | 2511 | |||
| AY360464.1 | - | 8 | 849 | |||
| BC038400.1 | - | 10 | 1088 | |||
| BC109294.1 | - | 22 | 2418 | |||
| BC109295.1 | - | 21 | 2371 | |||

VWA3A
Genomic Map