siRNA Details
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034144.1 | 3 | 1780 | |||
NR_034145.1 | 4 | 1942 | ||||
GenBank mRNA | AK092009.1 | - | 4 | 1943 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034144.1 | 3 | 1780 | |||
NR_034145.1 | 4 | 1942 | ||||
GenBank mRNA | AK092009.1 | - | 4 | 1943 |