siRNA Details
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034145.1 | 2 | 171 | |||
GenBank mRNA | AK092009.1 | - | 2 | 170 | ||
DB255288.1 | - | 2 | 172 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034145.1 | 2 | 171 | |||
GenBank mRNA | AK092009.1 | - | 2 | 170 | ||
DB255288.1 | - | 2 | 172 |