siRNA Details
| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026974.1 | 1 | 1773 | |||
| GenBank mRNA | AK092777.1 | - | 1 | 1598 | ||
| BC046439.1 | - | 1 | 1773 | |||
| BC122544.1 | - | 1 | 1747 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF252P-AS1| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026974.1 | 1 | 1773 | |||
| GenBank mRNA | AK092777.1 | - | 1 | 1598 | ||
| BC046439.1 | - | 1 | 1773 | |||
| BC122544.1 | - | 1 | 1747 | |||