siRNA Details
| Gene Symbol: ZNF667-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036521.1 | 2 | 1670 | |||
| NR_036522.1 | 2 | 1315 | ||||
| GenBank mRNA | BC008651.1 | - | 3 | 552 | ||
| BC110411.1 | - | 2 | 1413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF667-AS1| Gene Symbol: ZNF667-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036521.1 | 2 | 1670 | |||
| NR_036522.1 | 2 | 1315 | ||||
| GenBank mRNA | BC008651.1 | - | 3 | 552 | ||
| BC110411.1 | - | 2 | 1413 | |||