siRNA Details
Gene Symbol: DKFZp779M0652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027134.1 | 2 | 625 | |||
GenBank mRNA | BX640618.1 | - | 2 | 625 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DKFZp779M0652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027134.1 | 2 | 625 | |||
GenBank mRNA | BX640618.1 | - | 2 | 625 |