siRNA Details
Gene Symbol: ILF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024333.1 | 1 | 1955 | |||
GenBank mRNA | AF439744.2 | - | 1 | 1565 | ||
AL832185.1 | - | 1 | 1929 | |||
BC044911.2 | - | 1 | 1931 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ILF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024333.1 | 1 | 1955 | |||
GenBank mRNA | AF439744.2 | - | 1 | 1565 | ||
AL832185.1 | - | 1 | 1929 | |||
BC044911.2 | - | 1 | 1931 |