siRNA Details
| Gene Symbol: DCLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033403.1 | NP_208382.1 | 2 | 167 | ||
| XM_011534167.2 | XP_011532469.1 | 2 | 126 | |||
| XM_011534168.2 | XP_011532470.1 | 2 | 167 | |||
| XM_011534169.1 | XP_011532471.1 | 2 | 167 | |||
| GenBank mRNA | DB075681.1 | - | 2 | 167 | ||

DCLK3
Genomic Map