siRNA Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288590.1 | NP_001275519.1 | 2 | 823 | ||
| NM_001288591.1 | NP_001275520.1 | 2 | 823 | |||
| NM_018651.3 | NP_061121.2 | 2 | 860 | |||
| NM_025169.2 | NP_079445.1 | 2 | 860 | |||
| GenBank mRNA | AF075093.1 | - | 1 | 458 | ||
| AF154846.1 | - | 1 | 476 | |||
| AK022800.1 | - | 2 | 769 | |||
| AK291285.1 | - | 2 | 805 | |||
| AY280798.1 | - | 2 | 814 | |||
| BC010555.1 | - | 2 | 778 | |||
| BC063020.1 | - | 2 | 812 | |||
| BC120982.1 | - | 2 | 552 | |||
| BC120983.1 | - | 2 | 591 | |||
| BC127639.1 | - | 2 | 591 | |||
| BC127640.1 | - | 2 | 591 | |||

ZKSCAN7
Genomic Map