siRNA Details
| Gene Symbol: USP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015276.1 | NP_056091.1 | 13 | 1836 | ||
| XM_005256575.2 | XP_005256632.1 | 13 | 1550 | |||
| GenBank mRNA | AB028986.1 | - | 13 | 1836 | ||
| BC007196.1 | - | 6 | 578 | |||
| BC025317.1 | - | 6 | 576 | |||
| BX640815.1 | - | 12 | 1418 | |||

USP22
Genomic Map