siRNA Details
| Gene Symbol: KIF26A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015656.1 | NP_056471.1 | 15 | 6333 | ||
| XM_011536641.2 | XP_011534943.1 | 12 | 5478 | |||
| GenBank mRNA | AB033062.2 | - | 14 | 6207 | ||
| AB290161.1 | - | 14 | 6303 | |||
| AL117589.1 | - | 4 | 1487 | |||
| BC009415.1 | - | 4 | 1521 | |||
| BC025353.1 | - | 4 | 2479 | |||
| BE540821.1 | - | 1 | 56 | |||

KIF26A
Genomic Map