siRNA Details
| Gene Symbol: ZNF395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018660.2 | NP_061130.1 | 2, 3 | 374 | ||
| GenBank mRNA | AB044750.1 | - | 1, 2 | 197 | ||
| AB073627.1 | - | 2, 3 | 380 | |||
| AF263928.1 | - | 3, 4 | 584 | |||
| AK002050.1 | - | 2, 3 | 299 | |||
| AK023418.1 | - | 2, 3 | 374 | |||
| AK098243.1 | - | 8, 9 | 1257 | |||
| BC001237.1 | - | 2, 3 | 336 | 3452822 | ||

ZNF395
Genomic Map