siRNA Details
| Gene Symbol: CCDC85A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080433.1 | NP_001073902.1 | 6 | 2536 | ||
| XM_005264125.1 | XP_005264182.1 | 5 | 1926 | |||
| GenBank mRNA | AB067499.2 | - | 6 | 2623 | ||
| BC131557.1 | - | 6 | 1208 | |||

CCDC85A
Genomic Map