siRNA Details
| Gene Symbol: LYSMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286812.1 | NP_001273741.1 | 3 | 566 | ||
| NM_198273.2 | NP_938014.1 | 3 | 625 | |||
| XM_005248421.4 | XP_005248478.1 | 3 | 1090 | |||
| GenBank mRNA | AK291478.1 | - | 3 | 622 | ||
| AL137315.1 | - | 1 | 122 | |||
| AL832721.1 | - | 3 | 606 | |||
| BC058027.1 | - | 3 | 566 | |||
| BC136740.1 | - | 3 | 517 | 9020730 | ||
| BC136741.1 | - | 3 | 517 | 9020731 | ||
| BX537972.1 | - | 3 | 595 | |||
| DN994993.1 | - | 3 | 634 | |||

LYSMD3
Genomic Map